L17905.06
L17905.03
L17905.06EA
290
EUR
InStock
L17905.06
L17905.03
Methyl-7-phenylheptanoate 98%
Methyl-7-phenylheptanoate
Purity: 98%
CAS No.: 101689-18-3
Molecular Formula: C6H5(CH2)6CO2CH3
Formula Weight: 220.31
Refractive Index: 1.4946
Boiling Point: ca 155°/1.5mm
Beilstein Reference: 2694776
MDL No.: MFCD02093971