Chromatography cartridges, glutathione, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
16109 16110
PIER16109EA 385 EUR InStock
PIER16109 PIER16110
Chromatography cartridges, glutathione, Pierce™
Chromatographiesäulen
Pierce™ Glutathione chromatography cartridges are convenient, ready to use pre-packed devices for the purification of GST-fusion proteins from cellular lysates. The glutathione is immobilised through its central sulfhydryl onto 6% crosslinked agarose resin. Purification of GST-fusion proteins using glutathione (GSH) agarose beads is well documented and provides a one-step, high purity affinity purification.
  • Binds at least 40 mg of purified recombinant GST protein per ml of resin
  • Consistently purifies at least 10 mg of GST-tagged protein per ml of resin with greater than 90% purity
  • Resin can be reused at least five times without reduction in binding capacity and purification performance
  • Works well to purify GST-fusion proteins from bacterial lysates or use with pre-purified GST-tagged proteins to pull down protein interactions
Pierce™ Glutathione chromatography cartridges are compatible with the major automated liquid-chromatography systems or manual syringe processing. The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-Lok® syringe fittings or 1/16" tubing. These cartridges enable fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
Lieferumfang: Kit includes an accessory pack (1 female Luer-Lok® adapter, 1 connector fitting, 1 column plug and bottom caps).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge