Accessories for Rapid Equilbrium Dialysis (RED) Device, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
89810 89809 90004 90005
PIER89810EA 2480 EUR InStock
PIER89810 PIER89809 PIER90004 PIER90005
Accessories for Rapid Equilbrium Dialysis (RED) Device, Pierce™
Röhrchen Dialyseröhrchen
These are single-use, disposable inserts made up of two side-by-side chambers separated by a vertical cylinder of dialysis membrane validated for minimal nonspecific binding.
  • Disposable inserts require no presoaking, assembly, or specialized equipment
  • Large dialysis surface area accelerates equilibrium giving rise to short incubation time
  • Ideal molecular-weight (8 kDa) cutoff for protein-drug binding studies
  • Regenerated cellulose with low glycerol content as a humectant
This requires no extensive assembly steps or specialsed equipment, and each chamber or well is easily accessible from the top of the insert after insertion in the base plate. Additionally, the high surface-to-volume ratio of the membrane compartment allows rapid dialysis, where equilibrium can be reached in 4 hours with high levels of reproducibility and accuracy. In many cases, experiments can be completed in less than 100 minutes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge