Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZAP70 Mouse Monoclonal Antibody (CF555) [clone: ZAP70/528 + 2F3.2]

Lieferant: Biotium

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

BNC551137-500 BNC551137-100
BNC551137-500EA 560 EUR InStock
BNC551137-500 BNC551137-100
Anti-ZAP70 Mouse Monoclonal Antibody (CF555) [clone: ZAP70/528 + 2F3.2]
Antikörper
ZAP70 is a 70 kDa protein tyrosine kinase found in T-cells and natural killer cells.Control of this protein translation is via the IgVH gene. ZAP70 protein is expressed in leukemic cells of approximately 25% of chronic lymphocytic leukemia (CLL) cases as well.Anti-ZAP70 expression is an excellent surrogate marker for the distinction between the Ig-mutated (anti-ZAP70 negative) and Ig-unmutated (anti-ZAP70 positive) CLL subtypes and can identify patient groups with divergent clinical courses. The anti-ZAP70 positive Ig-unmutated CLL cases have been shown to have a poorer prognosis.

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: ZAP70/528 + 2F3.2
Conjugation: CF555
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge