HPLC columns, Allure® organic acids
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
9165563-700 9165565-700 9165575-700 9165585-700 9165563 9165565 9165575 9165585 916550210 916550212 916550220 916550222 916556E
REST9165563-700EA 0 EUR InStock
REST9165563-700 REST9165565-700 REST9165575-700 REST9165585-700 REST9165563 REST9165565 REST9165575 REST9165585 REST916550210 REST916550212 REST916550220 REST916550222 REST916556E
HPLC columns, Allure® organic acids
Chromatographiesäulen
Allure® Organic Acids columns provide enhanced retention and selectivity for polar organic acids, allowing the separation to be performed on a single 30 cm column.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge