Indicating hydrocarbon traps for air compressors
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
20637 20636
REST20637EA 0 EUR InStock
REST20637 REST20636
Indicating hydrocarbon traps for air compressors
Filter Gasfilter
These indicating hydrocarbon traps for air compressors remove oil vapours and mist as it passes compressed air from an oil-filled air compressor. Indicator changes from pale pink to deep pink.
Dimensions: 152×44 mm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge