Moisture control by-pass lines
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
21035 21109
REST21035EA 48 EUR InStock
REST21035 REST21109
Moisture control by-pass lines
Schläuche Transferschlauch
These moisture control by-pass lines for Tekmar instruments are suitable for U.S EPA methods 8260, 524.2 and OLM4.1.
  • Silcosteel®-deactivated tubing for increased inertness
  • Increases response for ketones, alcohols and acetates
  • Easy attachment in minutes
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge