Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

High-performance Blood Bank Refrigerator, TSX

Lieferant: HERAEUS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

TSX2304BV TSX1204BV TSX3004BV TSX5004BV
HERATSX2304BVEA 8572 EUR InStock
HERATSX2304BV HERATSX1204BV HERATSX3004BV HERATSX5004BV
High-performance Blood Bank Refrigerator, TSX
Kühlschränke
Thermo Scientific™ TSX Series high-performance blood bank refrigerator is designed with features that support sample protection and sustainability objectives for the storage of whole blood and blood components. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

  • Factory pre-set to 4 °C to meet blood storage guidelines
  • Triple pane glass door helps maintain optimal internal condition
  • Forced-air circulation for temperature uniformity and fast temperature recovery
  • Heat-free defrost for maximum temperature uniformity
  • Standard, built-in chart recorder
  • Quiet operation at just 52 dBA
  • Keyed on/off switch with easy to use, setpoint security
  • Self-closing door with 90° stop to assist with inventory loads
  • Built-in remote alarm contacts

V-drive technology: While conventional refrigerant, high-performance refrigerators and freezers use single speed compressors that continually cycle on and off, the TSX Series V-drive runs at variable speeds to adjust cooling performance to the conditions inside and outside of the refrigerator or freezer. When combined with the automated tuning control, this variable speed drive optimizes the compressors running speed to the current conditions. When there are frequent door openings, or samples are added to the refrigerator or freezer, the control system detects the activity and increases the drive speed to bring temperatures back to the set point quickly. This innovative technology is one of the reasons the TSX Series delivers outstanding door opening recovery (DOR) speed, and more peace of mind for busy laboratories. When conditions are stable, such as when the unit is running overnight or on weekends, the drive runs at a low speed, reducing energy consumption while maintaining a stable temperature for the samples.

Zertifizierungen: Designed to meet strict requirements established bu the AABB for the storage or whole blood and blood components.
UL, cUL, CE.

Order Now
 

PRODUKT

TECHNISCHE DATEN

SERVICELEISTUNGEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge