B20587.06
B20587.14
B20587.06EA
32.2
EUR
InStock
B20587.06
B20587.14
2,2,3,4,4,4-Hexafluor-1-butanol 95%
2,2,3,4,4,4-Hexafluor-1-butanol
Purity: 95%
CAS No.: 382-31-0
Molecular Formula: CF3CHFCF2CH2OH
Formula Weight: 182.07
Flash Point: 51°(123°F)
Refractive Index: 1.3120
Boiling Point: 114-116°
Beilstein Reference: 1753872
UN No.: 1993
MDL No.: MFCD00042309