Replacement cap frit filters for Trident guard cartridges
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
25057 25022 25023 25990
REST25057EA 52 EUR InStock
REST25057 REST25022 REST25023 REST25990
Replacement cap frit filters for Trident guard cartridges
Filter Vorsäulen-Filter und -Fritten
The removable cap frit filters in a Trident help to prevent clogged cartridges and to extend the life of the column and cartridge.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge