31887
31887.15
31887.25
31888.15
31888.25
31889.15
31889.25
31888
31889
REST31887EA
66
EUR
InStock
REST31887
REST31887.15
REST31887.25
REST31888.15
REST31888.25
REST31889.15
REST31889.25
REST31888
REST31889
Revised B/N surrogate mix
Standards
Organische Standards
This mix contains four components (2-fluorobiphenyl, nitrobenzene-d5, p-terphenyl-d14, pyrene-d10).