Hydrophobic interaction chromatography columns, InfinityLab AdvanceBio
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
681975-908 685975-908
HEWL681975-908EA 0 EUR InStock
HEWL681975-908 HEWL685975-908
Hydrophobic interaction chromatography columns, InfinityLab AdvanceBio
Chromatographiesäulen
These columns are suitable for robust hydrophobic interaction chromatography of intact native proteins.
  • Faster method development and higher throughput for greater productivity
  • Single chemistry, multiple applications with better peak shape
  • Enhanced robustness, batch-to-batch consistency
Designed to address particularly challenging molecules, such as monoclonal antibodies (mAbs), antibody drug conjugates (ADCs), and other recombinant proteins.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge