A11923.09
A11923.18
A11923.30
A11923.09EA
44.1
EUR
InStock
A11923.09
A11923.18
A11923.30
1,3-Propansulton ≥99%
1,3-Propansulton
Purity: 99%
CAS No.: 1120-71-4
Molecular Formula: C3H6O3S
Formula Weight: 122.14
Flash Point: 110°(230°F)
Melting Point: 30-33°
Boiling Point: 179-181°/30mm
Beilstein Reference: 109782
UN No.: 2811
MDL No.: MFCD00005355