ESD Safety Sandals, Velcro®, Hannah XXTL Low
Lieferant: ELTEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
741531/35 741531/36 741531/37 741531/38 741531/39 741531/40 741531/41 741531/42
ELTN741531/35EA 90.9 EUR InStock
ELTN741531/35 ELTN741531/36 ELTN741531/37 ELTN741531/38 ELTN741531/39 ELTN741531/40 ELTN741531/41 ELTN741531/42
ESD Safety Sandals, Velcro®, Hannah XXTL Low
Schuhe und Stiefel
This leather-free ladies' safety sandal is suitable for work in ESD sensitive or electrostatically protected areas (EPA). It features a padded tongle, fabric lining and a PU/PU sole, Wellmax Trainers Lady.
  • Microfibre/textile material construction with fabric lining for natural ventilation
  • Full-length inlay sole, ESD PRO Lady Y
  • Non-metallic, penetration-resistant midsole
  • Steel toe cap
  • Sole core made of Infinergy® by BASF
Colour: Black/red
Zertifizierungen: EN ISO 20345 S1P SRC, form A
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge