DuraSleeve® Rigid Wire Marking Inserts for M710
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
312004 312006 311999 311996 312001
BRDY312004EA 46.5 EUR InStock
BRDY312004 BRDY312006 BRDY311999 BRDY311996 BRDY312001
DuraSleeve® Rigid Wire Marking Inserts for M710
Etiketten
The rigid construction of the DuraSleeve® wire marking system provides inserts that can easily be inserted into a clean carrier.
  • Brady DuraSleeve® printable inserts create a durable legend
  • The legend can be changed before or after termination
  • Matte finish
Applications include data and telecommunications labelling, electrical labelling, and wire and cable labelling.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge