BradyGrip® Print-on-Hook Labels with VELCRO® Hook for M710 and BMP71
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
173270 173238 173202 173299 173230
BRDY173270EA 89.13 EUR InStock
BRDY173270 BRDY173238 BRDY173202 BRDY173299 BRDY173230
BradyGrip® Print-on-Hook Labels with VELCRO® Hook for M710 and BMP71
Etiketten
For wire and cable ID, use BradyGrip® print-on hook material, an exclusive print-and-place material that can be easily applied, repositioned, or removed.
  • Features a printable front and grippy back with tiny hooks
  • Attaches to VELCRO® brand ONE-WRAP® strap for easy wire and cable ID
  • Will not damage wires and cables like nylon cable ties
  • Maximise uptime with faster, printable ID on cable bundles
  • Matte finish
Removable and repositionable are ideally suited for wire and cable labelling. A ribbon is required to print this material. Use part numbers M7-R6000 and M71-R6000.
VELCRO® brand ONE-WRAP® product not included with purchase. VELCRO® and ONE-WRAP® are registered trademarks of Velcro IP holdings LLC. Used with permission.
Verpackung: One tape per box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge