High Temp Flame Retardant Polyimide Wrap Around Labels for M710 and BMP71
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
174600 173343 173262 173261 173296 174557 173295 173268 173269
BRDY174600EA 139.6 EUR InStock
BRDY174600 BRDY173343 BRDY173262 BRDY173261 BRDY173296 BRDY174557 BRDY173295 BRDY173268 BRDY173269
High Temp Flame Retardant Polyimide Wrap Around Labels for M710 and BMP71
Etiketten
Flame retardant wire wraps (B-472) are ideal for cable and wire bundle applications and label applications where self-extinguishing properties are required.
  • Polyimide material specifically designed to self−extinguish and prevent flames from propagating
  • Matte finish material with permanent acrylic adhesive can be thermal transfer printed or dot matrix printed
  • Service temperature of −40°C to +100 °C (−40 to +212 °F)
Ideal for data and telecommunications labelling, electrical labelling, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R4300.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge