Ultra Aggressive Adhesive Metallised Matte Polyester Labels for M710 and BMP71
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
173327 173349 173379 173357 173406
BRDY173327EA 68.77 EUR InStock
BRDY173327 BRDY173349 BRDY173379 BRDY173357 BRDY173406
Ultra Aggressive Adhesive Metallised Matte Polyester Labels for M710 and BMP71
Etiketten
Ultra aggressive metallised polyester (B-486) is ideal for rating/serial plates that use barcodes, alphanumerics, graphic symbols and logos, and require nameplate-like quality.
  • Matte, light grey material has a permanent rubber-based adhesive and is thermal transfer printable
  • Designed for high adhesion to textured metals, powder-coated surfaces, and low energy plastics
  • Service temperature of −40 to +120 °C (−40 to +248 °F)
Ideal for asset tracking, component and equipment labelling, general identification, inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R4300.
Zertifizierungen: UL recognised. CSA accepted.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge