AlphaTec® 04-004 Food Processing Gloves
Lieferant: ANSELL HEALTH CARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
04-004/10 04-004/11 04-004/8 04-004/9
ANSE04-004/10EA 152 EUR InStock
ANSE04-004/10 ANSE04-004/11 ANSE04-004/8 ANSE04-004/9
AlphaTec® 04-004 Food Processing Gloves
Handschuhe
These gloves are robust, food handling-compliant PVC gloves, granting better chemical protection and grip.
  • Special nitrile and PVC compound makes it resistance to wide-ranging chemicals
  • Ideal food safety gloves for various applications
  • 13 gauge seamless cotton design providing supreme comfort and hand protection
These AlphaTec® gloves also feature a unique foam grip design that repels oil with ease, while ensuring excellent grip performance in wet and oily environments.
AlphaTec® 04-004 heavy-duty gloves boast an ergonomic hand shape that promotes wearer comfort and ease of use.
Recommended for: All food processing applications that have food contact, chemical handling, maintenance and cleaning.
Primary industries: Food processing, infrastructure maintenance and chemical.
Zertifizierungen: EN 420:2003 + A1:2009, EN ISO 374-1:2016 Type A, EN 388
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge