A15198.06
A15198.14
A15198.06EA
51.6
EUR
InStock
A15198.06
A15198.14
3-Brom-4-fluorbenzaldehyd ≥98%
3-Bromo-4-fluorobenzaldehyde
Purity: 98%
CAS No.: 77771-02-9
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Flash Point: 110°(230°F)
Refractive Index: 1.5740
Melting Point: 29-33°
Boiling Point: 138-139°/2.5mm
Beilstein Reference: 5806226
MDL No.: MFCD00042186