L01306.22
L01306.36
L01306.22EA
29.3
EUR
InStock
L01306.22
L01306.36
1-Phenyl-2-harnstoff ≥97%
1-Phenyl-2-harnstoff
Purity: 97%
CAS No.: 64-10-8
Molecular Formula: C7H8N2O
Formula Weight: 136.15
Melting Point: 144-148°
Boiling Point: 238°
Merck Reference: 14,7319
MDL No.: MFCD00007944