Stabilization:
Stabilisiert mit 4-tert-Butylbrenzcatechin (TBC) 1000 ppm
42804.14
42804.22
42804.36
42804.14EA
23.2
EUR
InStock
42804.14
42804.22
42804.36
Divinylbenzol (Isomerengemisch) ≥80% stabilisiert
Divinylbenzol (Isomerengemisch)
Stabilized with 1000 ppm 4-tert-butylcatechol
Purity: 80%
CAS No.: 1321-74-0
Molecular Formula: C6H4(CH=CH2)2
Formula Weight: 130.19
Storage Temperature: 2°C to 8°C
Flash Point: 69°(156°F)
Physical Form: Liquid
Density: 0.914
Refractive Index: 1.5740
Sensitivity: Light Sensitive
Appearance: Colourless to pale yellow
MDL No.: MFCD01778656