Pump tubing, Viton®
Lieferant: Heidolph Instruments GmbH & Co.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
525-50026-00 525-50027-00 525-50028-00 525-52000-00 525-53000-00 525-54000-00 525-56000-00
HEID525-50026-00EA 42 EUR InStock
HEID525-50026-00 HEID525-50027-00 HEID525-50028-00 224-1470 224-1466 224-1467 224-1469
Pump tubing, Viton®
Schläuche Transferschlauch
Fluorocarbon rubber, thermoformed Viton B (67% fluorinated), opaque black.
  • Low gas permeability
  • Resistant to solvents and corrosives at high temperatures
Excellent acid resistance at high temperatures.
Temperature range: -30 to +205 °C
Zertifizierungen: GLP conform.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge