L06696.14
L06696.06
L06696.22
L06696.14EA
46
EUR
InStock
L06696.14
L06696.06
L06696.22
1-Naphthylacetonitril ≥97%
1-Naphthylacetonitril
Purity: 97%
CAS No.: 132-75-2
Molecular Formula: C12H9N
Formula Weight: 167.21
Flash Point: 110°(230°F)
Refractive Index: 1.6192
Melting Point: 33-35°
Boiling Point: 180-182°/9mm
Beilstein Reference: 1101012
UN No.: 3439
MDL No.: MFCD00004041