Drucken

Dimethylsulfoxid, SupraSolv® GC_HS, Supelco®

Lieferant: MERCK MILLIPORE
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5


MQ100

1.01900.1000 1.01900.2500 1.01900.0500
1.01900.1000EA 139.36 EUR
1.01900.1000 1.01900.2500 1.01900.0500
Dimethylsulfoxid, SupraSolv® GC_HS, Supelco®
Dimethylsulfoxid

SupraSolv® solvents for headspace gas chromatography are developed particularly for the analysis of residual
solvents in drug substances, excipients and drug products according to ICH, Ph. Eur. and USP - SupraSolv® headspace solvents are tested for all residual solvents defined by the ICH.

Formel: (CH₃)₂SO
MW: 78.14 g/mol
Siedepunkt: 189
Melting Pt: 18.5
Dichte: 1.101 g/cm³ (20 °C)
Flash-Pt: 87
Storage Temperature: Raumtemperatur
MDL Number: MFCD00002089
CAS-Nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=d0727d3220885f6adab5f099d9fd3b4de38d5710&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Solubility 1000 g/L (20 °C)
Molar Mass 18.4 °C
Bulk Density 78.1 g/mol
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.48 (20 °C)
Explosion Limit 1.8 - 63.0%(V)
Density 1.10 g/cm³ (20 °C)
Ignition Point 301 °C
Saturation Concentration 8.0 g/m³ (20 °C)
Viscosity Kinematic 2.14 mm²/s


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR