Drucken

Zink, Certipur® Reag. Ph. Eur. Standard_volumetric_secondary_material, Complexometry traceable to NIST, Supelco®

Lieferant: MERCK MILLIPORE
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Achtung


MQ100

1.02409.0100 1.02409.0103
1.02409.0100EA 139 EUR
1.02409.0100 1.02409.0103
Zink, Certipur® Reag. Ph. Eur. Standard_volumetric_secondary_material, Complexometry traceable to NIST, Supelco®
Zinc

Merck have introduced into the range of Certipur® standards some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: Zn
MW: 65.39 g/mol
Siedepunkt: 906 °C (1013 hPa)
Melting Pt: 419 °C
Dichte: 7.14 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011291
CAS-Nummer: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9 III
Merck Index: 12,10255

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Melting Point 419 °C
Molar Mass 65.39 g/mol
Boiling Point 906 °C (1013 hPa)
Density 7.13 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR