Drucken

(±)-2-Methyltetrahydrofuran, EMPLURA®

Lieferant: MERCK MILLIPORE
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Gefahr


MQ200

1.08292.2500 1.08292.1000 1.08292.4000
1.08292.2500EA 96.8 EUR
1.08292.2500 1.08292.1000 1.08292.4000
(±)-2-Methyltetrahydrofuran, EMPLURA®
(±)-2-Methyltetrahydrofuran

2-Methyltetrahydrofuran (Methyl THF) is a safer and more sustainable alternative to dichloromethane and tetrahydrofuran. It is derived from renewable resources, such as corncobs and sugarcane bagasse.


  • Less solvent consumption due to more efficient extraction and higher reaction yields
  • Lower volatility and higher flash point increase user safety
  • Limited miscibility in water reduces waste stream
  • Reliable availability (independent of petrochemical production)

Formel: C₅H₁₀O
MW: 86.13 g/mol
Siedepunkt: 79
Melting Pt: -136
Flash-Pt: -12
Storage Temperature: Raumtemperatur
MDL Number: MFCD00005367
CAS-Nummer: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3 II

Order Now

Spezifikation Testergebnisse

Hill Formula C₅H₁₀O
Molar Mass 86.13 g/mol
Boiling point 78 - 80 °C
Density 0.85 g/cm³ (20 °C)
Explosion limit 1.2 - 5.7 %(V)
Flash point -12 °C
Ignition temperature 270 °C
Melting Point -136 °C
Vapor pressure 130 hPa (25 °C)
Solubility 130 g/l (25 °C)
LD 50 oral LD50 Rat300 - 2000 mg/kg
LD 50 dermal LD50 Rat2000 mg/kg
Storage class 3 Flammable liquids
WGK WGK 2 water endangering
R Phrase R 11-19-22-38-41
S Phrase S 16-29-33-26-39


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR