Drucken

Hanus-Lösung zur Bestimmung der Iodzahl c(Ibr) = 0,1 mol/l Titripur® c(IBr) = 0.1 mol/l, Titripur® Iodine_value, Supelco®

Lieferant: MERCK MILLIPORE
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Gefahr


MQ100

1.09164.1000 1.09164.1003
1.09164.1000EA 142 EUR
1.09164.1000 1.09164.1003
Hanus-Lösung zur Bestimmung der Iodzahl c(Ibr) = 0,1 mol/l Titripur® c(IBr) = 0.1 mol/l, Titripur® Iodine_value, Supelco®
Hanus solution (Iodine bromine)

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: IBr
Dichte: 1.06 g/cm3 (20 °C)
Flash-Pt: 40 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011353
UN: 2789
ADR: 8 (3) II

Order Now

Spezifikation Testergebnisse

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR