DNA transfection kit for protein production, FectoCHO® Expression system
Lieferant: POLYPLUS TRANSFECTIONGesamtbewertung: 0
Bewertung: von 5
FectoCHO® expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD expression medium and a powerful transfection reagent, FectoPRO®.
- Outstanding protein production yields in various CHO cells
- Fast and effortless CHO cell adaptation
- Chemically defined kit components
- Excellent lot to lot reproducibility
- Cost-effective solution
FectoCHO® CD expression medium allows easy cultivation of various strains of CHO cells such as CHO-K1, CHO-S and ExpiCHO®-S , without the need of an extensive sequential adaptation. FectoPRO®-mediated transient transfection in this chemically defined medium leads to the highest protein and antibody yields ever reached using Transient Gene Expression, up to 4 times higher than with similar systems.
Zertifizierungen: Das Unternehmen ist seit 2002 entsprechend der ISO 9001 zertifiziert. Seitdem wird die Zertifizierung alle drei Jahre erneuert.
Bestellinformation: FectoPRO® transfection reagent and FectoCHO® CD expression medium can be purchased separately.
Lieferumfang: Jedes Kit enthält zu gleichen Teilen FectoPRO-Reagenz und FectoPRO™-Booster. FectoPRO™-Booster ist ebenfalls einzeln erhältlich.