Drucken
Nylon probe tools, Menda
Lieferant: DESCO
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
35619 35621 35622
115-3064EA 2.8 EUR
115-3064 115-3065 115-3066
Nylon probe tools, Menda
Werkzeugsätze
Ideal for opening plastic cases such as MP3 players, cell phones, laptops and more. Also a great tool for popping out batteries.
  • Nylon finish will not mark delicate surfaces
  • May be suitable for use in ESD protected areas
  • Temperature resistant