Drucken
Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Lieferant: VWR Seradigm
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
SERD76419-586EA 71.2 EUR
SERD76419-586 SERD76419-590 1300-500CR 1300-500CG 1300-500CGH 1300-500CH
Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Seren
Avantor Seradigm Select Grade FBS provides superior performance and consistency in cost-sensitive applications.
  • Industry-leading collection techniques at the source
  • Risk mitigation through a robust quality system
  • Proven track record for performance and consistency
  • Sourced from countries with health status classified as “Negligible BSE Risk” by the World Health Organization
The care and attention we devote to the collection and manufacture of our products generates quality that matters. Avantor Seradigm Select Grade FBS preserves more of the naturally-occurring growth factors that cells require to thrive, resulting in the highest possible quality sera for a price that fits your budget.
Example Certificates of Analysis show typical results. Select Grade FBS is sourced from countries in Central America.