Drucken

Size exclusion chromatography columns, illustra™ NAP™

Lieferant: Cytiva
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

17-0852-01 17-0852-02 17-0853-01 17-0853-02 17-0854-01 17-0854-02
17-0852-01EA 294 EUR
17-0852-01 17-0852-02 17-0853-01 17-0853-02 17-0854-01 17-0854-02
Size exclusion chromatography columns, illustra™ NAP™
Chromatographiesäulen
NAP™ columns are disposable columns prepacked with Sephadex™ G-25 DNA grade and require only gravity to run.

  • For rapid and efficient purification of DNA and oligonucleotides (>10-mers) in less than 15 minutes by gravity flow
  • Prepacked with Sephadex™ G-25 DNA grade in distilled water containing 0,15% Kathon CG/ICP Biocide
  • Available in three sizes depending on sample volume: 0,5 ml (NAP™-5), 1 ml (NAP™-10) or 2,5 ml (NAP™-25)
  • Useful for small-scale purification, desalting and buffer exchange

Lieferumfang: Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration, washing and sample elution.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR