Drucken
Multimodal chromatography columns, HiTrap™ Capto Core 400
Lieferant: Cytiva
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
17-3724-11
17-3724-11EA 467 EUR
17-3724-11
Multimodal chromatography columns, HiTrap™ Capto Core 400
Chromatographiesäulen
Novel core bead technology and multimodal ligand give Capto Core 400 dual functionality, size exclusion and binding chromatography, in one chromatography resin.
  • High capacity and productivity in flowthrough mode
  • Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification
  • Wide operational window of pH and conductivity
Capto Core 400 is an excellent alternative to size‑exclusion chromatography (SEC) resins that are typically employed in the final stages of virus purification in vaccine manufacture.