Drucken
Hydrophobic interaction chromatography media, Capto™ Butyl
Lieferant: GE HEALTHCARE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
17-5459-01
17-5459-01EA 171 EUR
17-5459-01
Hydrophobic interaction chromatography media, Capto™ Butyl
Protein-Aufbereitungs Protein Purification Resins
Capto™ Butyl is used in the capture and intermediate stages of protein purification. The combination of high capacity, high flow rate and low back pressure reduces process cycle times.
  • Improved productivity and process economy in downstream operations
  • Very high flow rates and large sample volume processing
  • Excellent chemical stability