Drucken

Salzsäure 32%, AnalaR NORMAPUR® zA

Lieferant: VWR Chemicals
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Gefahr

20254.321EA 50.7 EUR
20254.321 20254.296 20254.310 20254.401 20254.360 20254.440
Salzsäure 32%, AnalaR NORMAPUR® zA
Salzsäure
Formel: HCl
MW: 36.46 g/mol
Melting Pt: –30 °C
Dichte: 1.18 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
REACH: 01-2119484862-27

Order Now

Spezifikation Testergebnisse

Assay 31.5 - 33.0 %
Colouration Max. 10 APHA
Density (20/4) 1.140 - 1.180
Free chlorine Max. 0.5 ppm
Ignition residue (SO4) Max. 5 ppm
Br (Bromide) Max. 50 ppm
NH4 (Ammonium) Max. 1 ppm
PO4 (Phosphate) Max. 0.5 ppm
SO3 (Sulphite) Max. 1 ppm
SO4 (Sulphate) Max. 1 ppm
Al (Aluminium) Max. 0.05 ppm
As (Arsenic) Max. 0.01 ppm
Ba (Barium) Max. 0.02 ppm
Be (Beryllium) Max. 0.02 ppm
Bi (Bismuth) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.04 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.2 ppm
Ge (Germanium) Max. 0.05 ppm
Hg (Mercury) Max. 0.01 ppm
K (Potassium) Max. 0.1 ppm
Li (Lithium) Max. 0.01 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.04 ppm
Pb (Lead) Max. 0.02 ppm
Sr (Strontium) Max. 0.01 ppm
Ti (Titanium) Max. 0.1 ppm
Tl (Thallium) Max. 0.05 ppm
V (Vanadium) Max. 0.01 ppm
Zn (Zinc) Max. 0.05 ppm
Zr (Zirconium) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR