Drucken
Dosing bottle, HDPE
Lieferant: SEMADENI
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
3020
215-0241EA 3.6 EUR
215-0241
Dosing bottle, HDPE
Flaschen Tropfflaschen
Natural or translucent cylindrical shaped dosage bottle with hinged cap, protecting the jet orifice of Ø 3 mm.
  • For easy handling and flow dosing
  • Waterproof bottle when cap is closed
  • Good chemical and temperature resistance