Drucken
MEM Nonessential amino acids, Corning®
Lieferant: MEDIATECH
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
25-025-CI
25-025-CIEA 121.58 EUR
25-025-CI
MEM Nonessential amino acids, Corning®
Zellkultur-Ergänzungen und -Zusätze
The addition of MEM nonessential amino acids to medium can improve growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells.
This concentrated supplement allows for the efficient addition to media with no risk of dilution of media components.