Drucken
HEPES Pufferlösung (1 mol/L), Corning®
Lieferant: MEDIATECH
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
25-060-CI
25-060-CIEA 0 EUR
25-060-CI
HEPES Pufferlösung (1 mol/L), Corning®
Puffer Gewebekultur-Puffer
HEPES (N-2-hydroxyethylpiperazine-N'-2-ethanesulphonic acid) provides an alternative to the use of bicarbonate for buffering because it does not require elevated levels of CO2
Unlike the low pKa of bicarbonate, HEPES has a pKa of 7.66 making it a much stronger buffer in the pH 7.2-7.6 range than bicarbonate buffer. Potential disadvantages of HEPES include toxicity over 100 mM and lack of nutritive value to the cell.