Drucken
Rat IgG2a Isotype Control [clone: 2A3] (APC (Allophycocyanin)-Cy7®)
Lieferant: Tonbo Biosciences
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
25-4321-U100
25-4321-U100EA 159 EUR
25-4321-U100
Rat IgG2a Isotype Control [clone: 2A3] (APC (Allophycocyanin)-Cy7®)
Antikörper
The A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin binds to TNP (trinitrophenol), a hapten not expressed on mouse or human cells. It is used as an isotype control for rat IgG2a antibodies.
Type: Control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: 2A3
Conjugation: APC-Cyanine7
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: N/A