Drucken
dNTP sets, illustra™
Lieferant: GE HEALTHCARE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
28-4065-51 28-4065-57 28-4065-52 28-4065-53 28-4065-58
28-4065-51EA 266 EUR
28-4065-51 28-4065-57 28-4065-52 28-4065-53 28-4065-58
dNTP sets, illustra™
Nukleinsäure-Reagenzien Nukleotide
High purity dNTPs for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for consistent, high performance
  • Buffer-free, ready to use solutions at a variety of concentrations
  • Functionally tested to produce a 20,7 kb PCR amplification product from lambda DNA