Drucken
Storage phosphor screens and cassettes
Lieferant: Cytiva
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
28-9564-82 28-9564-78 28-9564-74 28-9564-75 28-9564-76 28-9564-77 28-9564-81 29-1755-23 29-1755-24
28-9564-82EA 1066 EUR
28-9564-82 28-9564-78 28-9564-74 28-9564-75 28-9564-76 28-9564-77 28-9564-81 29-1755-23 29-1755-24
Storage phosphor screens and cassettes
Autoradiographie-Filme Kassetten
Storage phosphor screens capture latent images produced by ionising radiation (X-ray, β, and γ emissions from isotopes such as 14C, 3H, 125I, 131I, 32P, 35S, 33P, and more.). Upon laser-induced stimulation, light is emitted from the storage phosphor screen in proportion to the amount of radioactivity in the sample. The resulting digital image allows for quantitation of subtle signal intensity differences over a wide dynamic range using storage phosphor imaging systems such as Typhoon, Storm, and PhosphorImager. Typically, 50% to 90% less exposure time is required compared with an equivalentMore Product Information