Drucken

Anti-IgG Goat Polyclonal Antibody (Biotin)

Lieferant: Tonbo Biosciences
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

30-8055-U500
30-8055-U500EA 188 EUR
30-8055-U500
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antikörper
Goat anti-Mouse IgG (H&L), F(ab')2 Fragment - Affinity Pure, Biotin Conjugate, min x w/human IgG or serum proteins

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR