Organic reference standard, Zearalenone-¹³C₁₈ solution, analytical standard, Supelco®
Lieferant: SUPELCO
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
32758-1ML
32758-1MLEA
2600
EUR
32758-1ML
Organic reference standard, Zearalenone-¹³C₁₈ solution, analytical standard, Supelco®
Standards
Organische Standards
Zearalenone-¹³C₁₈ may be used as an analytical standard for the determination of zearalenone in animal feed by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).
Zearalenone-¹³C₁₈ is the ¹³C isotope-labeled analog of the mycotoxin, zearalenone.