Drucken

Kaliumhydrogenphthalat 99,95-100,05% ACS primärer Standard

Lieferant: ALFA AESAR
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Achtung

33351.30 33351.A1
33351.30EA 68.5 EUR
33351.30 33351.A1
Kaliumhydrogenphthalat 99,95-100,05% ACS primärer Standard
Kaliumhydrogenphthalat

Purity: 99.95-100.05 CAS No.: 877-24-7 Molecular Formula: C8H5KO4 Formula Weight: 204.23 Physical Form: Crystalline powder Density: 1.636 Appearance: White Odour: Odourless Melting Point: 297° dec. Beilstein Reference: 3637128 Merck Reference: 14,7612 MDL No.: MFCD00013070


As a primary standard for preparing volumetric alkali solutions and as a buffer in pH determinations


Achtung! Bitte beachten: Incompatible with strong oxidising agents.

Formel: HOOCC₆H₄COOK
MW: 204.22 g/mol
Melting Pt: 297 °C (Zers.)
Dichte: 1.636
Storage Temperature: Raumtemperatur
MDL Number: MFCD00013070
CAS-Nummer: 877-24-7
EINECS: 212-889-4
Merck Index: 14,7612
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=cc8e011180c68e19e524dd3eb6f3b34f86486515&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR