Drucken

Lecithin, raffiniert

Lieferant: ALFA AESAR
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

36486.30 36486.A1
36486.30EA 74 EUR
36486.30 36486.A1
Lecithin, raffiniert
Lecithin

Lecithin is used as an emulsifying and stabilizing agent in the food, pharmaceutical, and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacturing of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.


CAS No.: 8002-43-5 Physical Form: Powder Odour: Odourless Beilstein Reference: 5209585 Merck Reference: 14,5428 MDL No.: MFCD00147406


Achtung! Bitte beachten: RUO – Research Use Only

Dichte: 1.0305 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00147406
CAS-Nummer: 8002-43-5
EINECS: 232-307-2
Merck Index: 14,5428

Order Now

Spezifikation Testergebnisse

Appearance (Color) Light tan to medium yellow
Form Powder
Assay (unspecified) Hexane insoluble 0.3%, max
Acid Value <35 mg KOH/g
Residual water < 1%
Peroxide content 5 meq /kg, max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR