Drucken

Racks and Boxes for Microcentrifuge Tubes

Lieferant: GLW
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

GSTY G50B G50G G50R VST
391-8855EA 3.6 EUR
391-8855 391-8856 391-8857 391-8858 GLWAVST
Racks and Boxes for Microcentrifuge Tubes
Boxen Boxen für Reagenzgläser
Styro racks are made of heavy expanded polystyrene which are used in freezer down to -90 °C. System box is made of cardboard and is designed to securely enclose two styro rack boxes.

  • Not autoclavable
  • For 1,5 and 2,0 ml tubes up to 12 mm diameter
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR