Drucken
VWR®, Cell culture media, Schneider's Drosophila medium
Lieferant: VWR Chemicals
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
392-0419EA 22.4 EUR
392-0419
VWR®, Cell culture media, Schneider's Drosophila medium
Zellkulturmedien
Schneider's Drosophila medium promotes rapid growth of both primary and existing cell cultures derived from Drosophila spp and several other dipterans when supplemented with 5 to 20% heat-inactivated foetal bovine serum.