Drucken
Anti-CD2 Rat Monoclonal Antibody (In Vivo Ready™) [clone: RM2-5]
Lieferant: Tonbo Biosciences
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
40-0021-U100
40-0021-U100EA 113 EUR
40-0021-U100
Anti-CD2 Rat Monoclonal Antibody (In Vivo Ready™) [clone: RM2-5]
Antikörper
The RM2-5 antibody reacts with mouse CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD48 in the mouse and is expressed on the cell surface of all lymphocytes. CD2 mediates cell to cell adhesion and is involved in T cell activation.
RM2-5 is reported to block CD2-mediated cell:cell adhesion. Please choose the appropriate format for each application.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RM2-5
Conjugation: In Vivo Ready™
Epitope:
Host: Rat
Isotype: IgG2b, lambda
Reactivity: Mouse