Drucken
Analytical mill, A 10 basic
Lieferant: IKA
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
4020700
412-0304EA 2333 EUR
412-0304
Analytical mill, A 10 basic
Mühlen
This batch mill has simple handling with keypads which grinds hard, brittle, soft and fibrous materials, because samples may be embrittled directly in the grinding chamber- tough, oily and aqueous samples can also be grinded. Easily exchangeable beater/cutter for a variety of applications.
  • Counter- displays grinding time and error code
  • Digital timer with interval function, integrated cooling connections
  • Brushless motor for long service life and low noise level
  • Bayonet lock for lid to enable simple and safe locking
The mill starts only when the lid is closed and can be opened at standstill. A quick stop feature further increases the safety of the user.