Phytochemical standard, Dihydromyricetin, analytical standard, Supelco®
Lieferant: SUPELCO
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
42866-10MG
42866-10MGEA
148
EUR
42866-10MG
Phytochemical standard, Dihydromyricetin, analytical standard, Supelco®
Standards
Organische Standards
Dihydromyricetin (ampelopsin) is a flavanonol with antioxidant and anti-cancer activity, found to have anti-alcohol intoxication effects. Its anti-alcohol effects appear to be by its actions as a positive modulator of GABA-A receptors at the benzodiazepine site.